Skip to Content
Merck
All Photos(2)

Key Documents

389471

Sigma-Aldrich

Ethyltriphenylphosphonium iodide

95%

Synonym(s):

ETPPI, Phenylphosphonium ethyl iodide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$146.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$146.00

About This Item

Linear Formula:
(C6H5)3P(I)C2H5
CAS Number:
Molecular Weight:
418.25
Beilstein:
3659323
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

164-168 °C (lit.)

functional group

phosphine

SMILES string

[I-].CC[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H20P.HI/c1-2-21(18-12-6-3-7-13-18,19-14-8-4-9-15-19)20-16-10-5-11-17-20;/h3-17H,2H2,1H3;1H/q+1;/p-1

InChI key

SLAFUPJSGFVWPP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ethyltriphenylphosphonium iodide can be used as:
  • An efficient catalyst for N,N-dimethylation of primary aromatic amines with methyl alkyl carbonates.[1]
  • A phase transfer catalyst for the synthesis of 3-hydroxyflavones from chalcones via Algar−Flynn−Oyamada synthesis.[2]

Reactant involved in:
  • Synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes
  • Asymmetric hydrogenation

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective N, N-dimethylation of primary aromatic amines with methyl alkyl carbonates in the presence of phosphonium salts
Selva M, et al.
The Journal of Organic Chemistry, 71(15), 5770-5773 (2006)
Phase Transfer Catalysis Extends The Scope of The Algar-Flynn-Oyamada Synthesis of 3-Hydroxyflavones
Nhu D, et al.
Australian Journal of Chemistry, 68(7), 1102-1107 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service