Skip to Content
Merck
All Photos(4)

Key Documents

189138

Sigma-Aldrich

Tetrabutylphosphonium bromide

greener alternative

98%

Synonym(s):

Tetra-n-butylphosphonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$158.00
500 G
$380.00

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$158.00
500 G
$380.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4P(Br)
CAS Number:
Molecular Weight:
339.33
Beilstein:
4160474
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

$158.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

100-103 °C (lit.)

greener alternative category

SMILES string

[Br-].CCCC[P+](CCCC)(CCCC)CCCC

InChI

1S/C16H36P.BrH/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;/h5-16H2,1-4H3;1H/q+1;/p-1

InChI key

RKHXQBLJXBGEKF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Tetrabutylphosphonium bromide (TBPB) is a quaternary salt which can be used:
  • As a medium to disperse ruthenium catalyst for the synthesis of ethylene glycol from synthesis gas via ruthenium melt catalysis.[1]
  • As a catalyst supported on silica or alumina for the halogen exchange reaction to synthesize alkyl bromide from alkyl chloride.[2]
  • To synthesize various ionic liquids on mixing with different proportions of 1,3-dimethylurea for capturing NO gas.[3]
  • As a hydrogen-bond acceptor along with levulinic acid as a hydrogen-bond donor for the preparation of deep eutectic solvents to separate toluene from toluene/n-hexane mixtures.[4]
  • To prepare ionic semiclathrate hydrates.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

554.0 °F - closed cup

Flash Point(C)

290 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 3-Dimethylurea tetrabutylphosphonium bromide ionic liquids for NO efficient and reversible capture
Jiang B, et al.
Energy and Fuels, 30(1), 735-739 (2016)
Yoshiro Yasaka et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 19(13), 1674-1682 (2018-04-25)
The mechanism of CO2 absorption by a formate ionic liquid, [P4444 ]HCOO, was studied by Raman spectroscopy. The band area for the symmetric CO2 stretching of the formate anion linearly decreases with the CO2 loading. From the slope of the
Thermodynamic properties of ionic semiclathrate hydrate formed with tetrabutylphosphonium bromide
Suginaka T
Fluid Phase Equilibria, 317(1), 25-28 (2012)
Separation of toluene from toluene/alkane mixtures with phosphonium salt based deep eutectic solvents
Hou Y et al.
Fuel Processing Technology, 135(1), 99-104 (2015)
Facile continuous process for gas phase halogen exchange over supported alkyl phosphonium salts
Sharma P and Sasson Y
Royal Society of Chemistry Advances, 8(5), 2824-2828 (2018)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service