Skip to Content
Merck
All Photos(1)

Key Documents

369519

Sigma-Aldrich

4,7,10-Trioxa-1,13-tridecanediamine

97%

Synonym(s):

O,O′-Bis(3-aminopropyl)diethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$78.20
500 G
$116.00

$78.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$78.20
500 G
$116.00

About This Item

Linear Formula:
O[CH2CH2O(CH2)3NH2]2
CAS Number:
Molecular Weight:
220.31
Beilstein:
1760709
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$78.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

146-148 °C/4 mmHg (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

NCCCOCCOCCOCCCN

InChI

1S/C10H24N2O3/c11-3-1-5-13-7-9-15-10-8-14-6-2-4-12/h1-12H2

InChI key

JCEZOHLWDIONSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,7,10-Trioxa-1,13-tridecanediamine can be used as a surface passivation agent for the synthesis of photoluminescent carbon nanodots for semiconductor applications.[1][2][3][4]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

282.2 °F

Flash Point(C)

139 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francisco Arriagada et al.
Nanomaterials (Basel, Switzerland), 9(2) (2019-02-10)
The design of efficient, biocompatible, and easily prepared vehicles for drug delivery is a subject of great interest for medicine and pharmaceutical sciences. To achieve the above goals, surface functionalization is critical. Here, we report a hybrid nanocarrier consisting of
One-step synthesis of surface passivated carbon nanodots by microwave assisted pyrolysis for enhanced multicolor photoluminescence and bioimaging
Liu C, et al.
Journal of Materials Chemistry, 21(35), 13163-13167 (2011)
Nano-carrier for gene delivery and bioimaging based on carbon dots with PEI-passivation enhanced fluorescence
Liu C, et al.
Biomaterials, 33(13), 3604-3613 (2012)
Highly luminescent carbon nanodots by microwave-assisted pyrolysis
Zhai X, et al.
Chemical Communications (Cambridge, England), 48(64), 7955-7957 (2012)
Fluorescent carbon nanodots conjugated with folic acid for distinguishing folate-receptor-positive cancer cells from normal cells
Song Y, et al.
Journal of Materials Chemistry, 22(25), 12568-12573 (2012)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service