Skip to Content
Merck
All Photos(1)

Key Documents

452572

Sigma-Aldrich

Poly(ethylene glycol) bis(3-aminopropyl) terminated

average MN 1,500, cross-linking reagent carboxyl reactive, amine

Synonym(s):

Polyethylene glycol, O,O′-Bis(3-aminopropyl)polyethylene glycol 1,500

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$92.50
5 G
$324.00

$92.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$92.50
5 G
$324.00

About This Item

Linear Formula:
(C2H4O)nC6H16N2O
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$92.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Poly(ethylene glycol) bis(3-aminopropyl) terminated, Mn ~1,500

mol wt

Mn ~1,500

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

mp

49 °C (lit.)

Ω-end

amine

α-end

amine

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

OCCO.NCCCO

InChI

1S/C3H9NO.C2H6O2/c4-2-1-3-5;3-1-2-4/h5H,1-4H2;3-4H,1-2H2

InChI key

ZEXFKFYUMFXUBE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization and preparation of PEG-polyimide copolymer asymmetric flat sheet membranes: This research discusses the preparation of asymmetric flat sheet membranes using Poly(ethylene glycol) bis(3-aminopropyl) terminated for carbon dioxide separation, enhancing the gas separation performance of the membranes (JH Park et al., 2015).


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of networked polymers by copolymerization of monoepoxy substituted lithium sulfonylimide.
Matsumoto K, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 49(8), 1874-1880 null
Khaled Habiba et al.
International journal of nanomedicine, 11, 107-119 (2016-01-15)
The combination of chemotherapy and photodynamic therapy has emerged as a promising strategy for cancer therapy due to its synergistic effects. In this work, PEGylated silver nanoparticles decorated with graphene quantum dots (Ag-GQDs) were tested as a platform to deliver
Liang-Chien Cheng et al.
Nanoscale, 5(9), 3931-3940 (2013-03-29)
This work demonstrates a simple route for synthesizing multi-functional fluorescent nanodiamond-gold/silver nanoparticles. The fluorescent nanodiamond is formed by the surface passivation of poly(ethylene glycol) bis(3-aminopropyl) terminated. Urchin-like gold/silver nanoparticles can be obtained via one-pot synthesis, and combined with each other

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the molecular weight of the polymer Poly(ethylene glycol), Product 452572?

    1 answer
    1. The molecular weight is approximately 1500.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service