Skip to Content
Merck
All Photos(2)

Documents

36816

Sigma-Aldrich

Diethylene glycol monoallyl ether

≥95.0% (GC)

Synonym(s):

Allyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
146.18
Beilstein:
1754585
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.446

density

1.010 g/mL at 20 °C (lit.)

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

C=CCOCCOCCO

InChI

1S/C7H14O3/c1-2-4-9-6-7-10-5-3-8/h2,8H,1,3-7H2

InChI key

DIOZVWSHACHNRT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D Fan et al.
Radiation protection dosimetry, 170(1-4), 311-314 (2016-03-05)
For more accurate assessments of the occupational exposure to radon for miners, the individual monitoring was conducted by using an improved passive integrating (222)Rn monitor. A total of 120 miners in 3 different kinds of mines were monitored throughout a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service