Skip to Content
Merck
All Photos(1)

Key Documents

499633

Sigma-Aldrich

Di(ethylene glycol) benzyl ether

97%

Synonym(s):

2-[2-(Benzyloxy)ethoxy]ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$167.00

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$167.00

About This Item

Linear Formula:
C6H5CH2OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
196.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.51 (lit.)

bp

80-110 °C/0.5 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl
phenyl

SMILES string

OCCOCCOCc1ccccc1

InChI

1S/C11H16O3/c12-6-7-13-8-9-14-10-11-4-2-1-3-5-11/h1-5,12H,6-10H2

InChI key

LJVNVNLFZQFJHU-UHFFFAOYSA-N

Application

Di(ethylene glycol) benzyl ether may be used as an initiator in the preparation of linear-PLA (poly(L-lactide).[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fei Wang et al.
Bioconjugate chemistry, 19(7), 1423-1429 (2008-06-21)
Two types of 32 arm star polymers incorporating amphiphilic block copolymer arms have been synthesized and characterized. The first type, stPCL-PEG 32, is composed of a polyamidoamine (PAMAM) dendrimer as the core with radiating arms having poly(epsilon-caprolactone) (PCL) as an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service