Skip to Content
Merck
All Photos(1)

Key Documents

366463

Sigma-Aldrich

Methyl nitroacetate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$552.00

$552.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$552.00

About This Item

Linear Formula:
NO2CH2COOCH3
CAS Number:
Molecular Weight:
119.08
Beilstein:
1758670
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$552.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

195-198 °C (lit.)

density

1.294 g/mL at 25 °C (lit.)

functional group

amine
ester
nitro

SMILES string

COC(=O)C[N+]([O-])=O

InChI

1S/C3H5NO4/c1-8-3(5)2-4(6)7/h2H2,1H3

InChI key

ALBSWLMUHHZLLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Methyl nitroacetate may be used:
  • in the preparation of aromatic and heteroaromatic linear (E)-α-nitro-arylpentenoates[1]
  • to generate phenyliodonium ylide, used in the highly enantioselective and diastereoselective Cu(I)-catalyzed cyclopropanation of alkenes[2]
  • in the preparation of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate, via reaction with 4-nitrobenzylideneaniline[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Andrew J Young et al.
Journal of the American Chemical Society, 130(43), 14090-14091 (2008-10-04)
The first electrophilic Pd(II)-catalyzed allylic C H alkylation is reported, providing a novel method for formation of sp3-sp3 C C bonds directly from C H bonds. A wide range of aromatic and heteroaromatic linear (E)-alpha-nitro-arylpentenoates are obtained as single olefin
Experimental and theoretical studies of Diels-Alder reaction between methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate and cyclopentadiene.
Jasinski R, et al.
Monatshefte fur Chemie / Chemical Monthly, 144(3), 327-335 (2013)
Benoît Moreau et al.
Journal of the American Chemical Society, 127(51), 18014-18015 (2005-12-22)
A highly enantioselective (up to 97.5% ee) and diastereoselective (95:5 dr trans/cis) Cu(I)-catalyzed cyclopropanation of alkenes using phenyliodonium ylide generated in situ from iodosobenzene and methyl nitroacetate is reported. The cyclopropanation took place with high enantioselectivity for a wide range
[Experimental establishment of the maximum permissible concentration of the methyl and ethyl esters of nitroacetic acid].
G I Sidorin et al.
Gigiena truda i professional'nye zabolevaniia, (7)(7), 49-51 (1980-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service