Skip to Content
Merck
All Photos(2)

Key Documents

227846

Sigma-Aldrich

Methyl 4-nitrobutyrate

97%

Synonym(s):

4-Nitrobutyric acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$190.00
25 G
$767.00

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$190.00
25 G
$767.00

About This Item

Linear Formula:
O2N(CH2)3CO2CH3
CAS Number:
Molecular Weight:
147.13
Beilstein:
1766154
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

106-110 °C/9 mmHg (lit.)

density

1.149 g/mL at 25 °C (lit.)

functional group

amine
ester
nitro

SMILES string

COC(=O)CCC[N+]([O-])=O

InChI

1S/C5H9NO4/c1-10-5(7)3-2-4-6(8)9/h2-4H2,1H3

InChI key

UBSPKGKFFQKZJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The reaction of methyl 4-nitrobutyrate with aldehydes, following the enantioselective Henry addition, was studied[1].

Other Notes

Remainder methyl 4-nitrovalerate

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gonzalo Blay et al.
Organic letters, 12(13), 3058-3061 (2010-06-12)
A catalytic highly enantioselective Henry addition of methyl 4-nitrobutyrate to aldehydes using a Cu(II)-amino pyridine complex as catalyst is described. The products resulting from this reaction constitute a new, highly versatile family of chiral building blocks as a result of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service