Merck
  • Calibration and field application of a molecularly imprinted membrane-passive sampler for the sampling of indicator polychlorinated biphenyls in selected aquatic environments of South Africa.
Calibration and field application of a molecularly imprinted membrane-passive sampler for the sampling of indicator polychlorinated biphenyls in selected aquatic environments of South Africa.
Water science and technology : a journal of the International Association on Water Pollution Research (2019-04-27)
Dennis S Mkhize, Laura P Quinn, Hlengilizwe Nyoni, Bhekie B Mamba, Titus A M Msagati
ABSTRACT
A passive sampling device, based on molecularly imprinted membranes (MIM), was fabricated and optimised for sampling polychlorinated biphenyls (PCBs) in aquatic ecosystems. The newly-developed passive sampler was subjected to in-situ calibration studies to determine PCB sampling rates under various conditions of water turbulence and temperature. This was carried out by exposing the passive samplers to water spiked with PCBs in a continuous-flow exposure setup. The samplers were preloaded with known concentrations of performance reference compounds (PRCs) prior to exposure. Sampling rates of seven indicator PCBs' congeners (PCBs 28, 52, 101, 118, 138, 153 and 180) ranged between 15.3 and 95.6 L/d for the different environmental conditions investigated. To determine the field suitability, the samplers were preloaded with PRCs and deployed for 10 days at the Roodeplaat and Hartbeespoort Dams, in South Africa. Water samples were taken at the end of the deployment period to compare the spot-and-grab samples to the developed samplers. PCBs 28, 101 and 138 were detected in the samplers deployed at Hartbeespoort Dam. The samplers deployed at the Roodeplaat Dam had quantifiable amounts of PCBs 28, 52, 101, 138 and 180 (0.047-0.828 ng mL-1 d-1). The sampler enhanced the detectability of PCB 52 and PCB 180, which were not detected in water samples. The field suitability trials indicated that the developed sampler could successfully be used for PCB monitoring. The sampler enhanced the detection of PCBs that would otherwise be too low to detect in samples collected through the traditional spot-and-grab sampling technique.