Skip to Content
Merck
All Photos(1)

Key Documents

364533

Sigma-Aldrich

Fluorotrimethylsilane

96%

Synonym(s):

Trimethylfluorosilane, Trimethylsilyl fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiF
CAS Number:
Molecular Weight:
92.19
Beilstein:
1731132
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

96%

bp

16 °C (lit.)

mp

−74 °C (lit.)

density

0.793 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)F

InChI

1S/C3H9FSi/c1-5(2,3)4/h1-3H3

InChI key

CTIKAHQFRQTTAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 1 - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

-22.0 °F - closed cup

Flash Point(C)

-30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L G Hutchins et al.
The International journal of applied radiation and isotopes, 36(5), 375-378 (1985-05-01)
18F-Labeled fluorotrimethylsilane was prepared by nucleophilic substitution of chlorotrimethylsilane with reactor produced [18F]fluoride. Hydrolysis of fluorotrimethylsilane by aqueous tetraethylammonium hydroxide followed by removal of water with a mechanical pump gave a powerful source of no carrier added nucleophilic 18F. Reaction
[Gas chromatographic fluoride determination following adsorption of trimethylfluorsilane on Porapak P].
K Seifert
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete, 29(2), 80-80 (1983-02-01)
M S Rosenthal et al.
The International journal of applied radiation and isotopes, 36(4), 318-319 (1985-04-01)
[18F]Fluorotrimethylsilane was prepared in 80% decay corrected yield by reaction of no-carrier added tetramethyl-ammonium fluoride (hydroxide as bulk anion) with chlorotrimethylsilane in 65% aqueous acetonitrile. The 18F gas was collected in a cold-trap at -130 degrees C and found to
Ian H Krouse et al.
Journal of the American Society for Mass Spectrometry, 16(5), 697-707 (2005-05-03)
In this study, preparation and decomposition of five novel pentavalent fluorosiliconates, RSi(CH3)3F- (R = CH3CH2O, CF3CH2O, (CH3)2CHO, (CH3)3SiO, and (CH3)3SiNH) is used to investigate the process of fluoride-induced desilylation. The siliconates were characterized by collision-induced dissociation and energy-resolved mass spectrometry.
P Venkateswarlu
Analytical chemistry, 64(4), 346-349 (1992-02-15)
The conventional procedure for separation of fluoride as trimethylfluorosilane in Conway diffusion cells involves the use of grease for sealing the cell and also for closing the hole in the lid drilled for introduction of hexamethyldisiloxane. We have developed a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service