Skip to Content
Merck
All Photos(1)

Key Documents

359912

Sigma-Aldrich

Benzyltriphenylphosphonium bromide + Sodium amide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$288.00

$288.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$288.00

About This Item

MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$288.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

258 °C (dec.) (lit.)

functional group

phenyl
phosphine

storage temp.

2-8°C

SMILES string

[Br-].N[Na].C(c1ccccc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C25H22P.BrH.H2N.Na/c1-5-13-22(14-6-1)21-26(23-15-7-2-8-16-23,24-17-9-3-10-18-24)25-19-11-4-12-20-25;;;/h1-20H,21H2;1H;1H2;/q+1;;-1;+1/p-1

InChI key

HEHHWWBQEVMFQU-UHFFFAOYSA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service