Skip to Content
Merck
All Photos(2)

Key Documents

270946

Sigma-Aldrich

Triphenylphosphine dibromide

96%

Synonym(s):

Bromotriphenylphosphonium bromide, Dibromotriphenylphosphorane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$52.00
25 G
$191.00

$52.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$52.00
25 G
$191.00

About This Item

Linear Formula:
(C6H5)3PBr2
CAS Number:
Molecular Weight:
422.09
Beilstein:
917652
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$52.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

96%

form

solid

mp

235 °C (dec.) (lit.)

functional group

phosphine

SMILES string

BrP(Br)(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C18H15Br2P/c19-21(20,16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

OCXGTPDKNBIOTF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used in the preparation of alkyl and aryl bromides from alcohols[1][2] and phenols[3] without rearrangement.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 22, 2945-2945 (1992)
Y Satoh et al.
Journal of medicinal chemistry, 36(23), 3580-3594 (1993-11-12)
A series of chromene derivatives was synthesized and evaluated for their in vitro and ex vivo 5-lipoxygenase (5-LO) inhibitory activity. These compounds were prepared by condensation of appropriate salicyl aldehydes with alpha, beta-unsaturated carbonyl compounds, followed by transformation to the
Christopher M Waldmann et al.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging, 20(2), 205-212 (2017-09-15)
The aim of this study was the automated synthesis of the mitochondrial membrane potential sensor 4-[18F]fluorobenzyl-triphenylphosphonium ([18F]FBnTP) on a commercially available synthesizer in activity yields (AY) that allow for imaging of multiple patients. A three-pot, four-step synthesis was implemented on
Journal of the American Chemical Society, 86, 964-964 (1964)
David Fuchs et al.
Journal of lipid research, 59(10), 2025-2033 (2018-08-02)
Trihydroxyoctadecenoic acids (TriHOMEs) are linoleic acid-derived oxylipins with potential physiological relevance in inflammatory processes as well as in maintaining an intact skin barrier. Due to the high number of possible TriHOME isomers with only subtle differences in their physicochemical properties

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service