Skip to Content
Merck
All Photos(3)

Key Documents

359084

Sigma-Aldrich

Sodium hydrogencitrate sesquihydrate

ReagentPlus®, 99%

Synonym(s):

Citric acid disodium salt, Citric acid disodium salt sesquihydrate, Di-Sodium hydrogen citrate sesquihydrate, Disodium hydrogen citrate sesquihydrate, Sodium citrate dibasic sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COONa)2 · 1.5H2O
CAS Number:
Molecular Weight:
263.11
Beilstein:
3770069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
99%

Quality Level

product line

ReagentPlus®

Assay

99%

form

solid

mp

149 °C (dec.) (lit.)

functional group

carboxylic acid

SMILES string

O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O.OC(=O)C(O)(CC([O-])=O)CC([O-])=O

InChI

1S/2C6H8O7.4Na.3H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;;3*1H2/q;;4*+1;;;/p-4

InChI key

HGPVLOQNBSHYEI-UHFFFAOYSA-J

Application

Sodium hydrogen citrate sesquihydrate is suitable reagent to investigate the mediatorless laccase cathodes as half-cell electrode under galvanostatic load by using a two-compartment set up. It may be used in the extraction of organ/tissue sample using QuEChERS (Quick, Easy, Cheap, Effective, Rugged, and Safe) extraction methods. It is suitable for use in the analysis of multiclass pesticides by multiresidue method based on the sample preparation by modified QuEChERS and detection by GC coupled to single quadruple mass spectrometery.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianna Marangi et al.
PloS one, 7(2), e31795-e31795 (2012-03-01)
In the poultry industry, control of the red mite D. gallinae primarily relies worldwide on acaricides registered for use in agriculture or for livestock, and those most widely used are carbamates, followed by amidines, pyrethroids and organophosphates. Due to the
Stefanie Rubenwolf et al.
Biosensors & bioelectronics, 26(2), 841-845 (2010-07-16)
Direct electron transfer from carbon electrodes to adsorbed laccase (EC 1.10.3.2) from Trametes versicolor is widely used to enable mediatorless enzymatic biofuel cell cathodes. However, data published so far are poorly comparable in terms of oxygen reduction performance. We thus
Jonas da Silva Sousa et al.
Food chemistry, 141(3), 2675-2681 (2013-07-23)
A multiresidue method based on the sample preparation by modified QuEChERS and detection by gas chromatography coupled to single quadruple mass spectrometers (GC-SQ/MS) was used for the analysis of 35 multiclass pesticides in melons (Cucumis melo inodorus) produced in Ceara-Brazil.
Mudasir Bhat et al.
Environmental monitoring and assessment, 187(7), 398-398 (2015-06-04)
Dissipation patterns were studied following two applications of difenoconazole (score 25% EC) at 300 and 600 g ai ha(-1) as single and double dose respectively on Golden Delicious and Starkrimson cultivars of apple. Samples were collected at 0, 3, 6
Malik Mukhtar et al.
Environmental monitoring and assessment, 187(8), 482-482 (2015-07-02)
The present studies were carried out to observe the dissipation pattern of chlorpyrifos on apple in Kashmir Valley. Persistence of chlorpyrifos in apple was studied following two applications rates of chlorpyrifos (Dursban 20 EC) at 200 g a.i. ha(-1) (single dose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service