Skip to Content
Merck
All Photos(1)

Key Documents

71497

Sigma-Aldrich

Sodium citrate monobasic

BioXtra, anhydrous, ≥99.5% (T)

Synonym(s):

Citric acid monosodium salt, Sodium dihydrogen citrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$112.00
1 KG
$378.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$112.00
1 KG
$378.00

About This Item

Linear Formula:
HOC(COONa)(CH2COOH)2
CAS Number:
Molecular Weight:
214.11
Beilstein:
3920246
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
crystals
solubility:
H2O: 0.25 M at 20 °C, clear, colorless

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

product line

BioXtra

Assay

≥99.5% (T)

form

crystals

impurities

insoluble matter, passes filter test
≤0.2% water

pH

3.5-4.0 (25 °C, 0.25 M in H2O)

solubility

H2O: 0.25 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.25 M in H2O

SMILES string

[Na+].OC(=O)CC(O)(CC([O-])=O)C(O)=O

InChI

1S/C6H8O7.Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);/q;+1/p-1

InChI key

HWPKGOGLCKPRLZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium citrate monobasic has been used-
  • as a component of Beltsville thawing solution (BTS) for semen dilution[1]
  • for the formulation of isonitrogenous diets to study the effects of sodium citrate in red hybrid tilapia[2]
  • for the preparation of SSC (saline sodium citrate) made for post-hybridization wash[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jarol R Miranda-Andrades et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 172, 126-134 (2016-05-01)
Gentamicin is an antibiotic indicated to treat mastitis in dairy cattle and for the treatment of bacterial resistance in the context of hospital infections. The effect caused by gentamicin on the optical properties of gold nanoparticles aqueous dispersions were used
M Solís et al.
Research in veterinary science, 83(3), 403-409 (2007-03-21)
Porcine rubulavirus (PoRV), also known as blue eye disease (BED) of swine, causes respiratory and reproductive problems in pigs at several developmental stages. To study the effect of PoRV infection on semen production, five boars were infected with 1 x
Dietary sodium citrate improved oxidative stability in red hybrid tilapia (Oreochromis sp.) but reduced growth, health status, intestinal short chain fatty acids and induced liver damage.
Romano N, et al.
Aquaculture (Amsterdam, Netherlands), 458, 170-176 (2016)
Jiayi Wang et al.
Food chemistry, 245, 337-345 (2017-12-31)
A new micro-plate colorimetric assay was developed for rapid determination of zinc in animal feed, pet food and drinking water. Zinc ion was extracted from sample by trichloroacetic acid and then reacted with 2-(5-Bromo-2-pyridylazo)-5-[N-propyl-N-(3-sulfopropyl)amino]phenol (5-Br-PAPS) to form a Zn-PAPS complex
Don-On Daniel Mak et al.
Cold Spring Harbor protocols, 2013(9), 880-884 (2013-09-05)
Nuclear patch-clamp experiments can be performed with intact nuclei or with nuclei from which the outer nuclear membrane has been removed. This protocol presents procedures for harvesting different types of cultured cells, isolating nuclei, and exposing the inner nuclear membrane

Articles

Highlighting existing and novel fabrication methods for both, solid and hydrogel-based scaffold for tissue engineering applications.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service