Merck
  • Degradation of chlorpyrifos residues in apple under temperate conditions of Kashmir Valley.
Degradation of chlorpyrifos residues in apple under temperate conditions of Kashmir Valley.
Environmental monitoring and assessment (2015-07-02)
Malik Mukhtar, Asma Sherwani, Ashraf Alam Wani, Sheikh Bilal Ahmed, Javid Ahmad Sofi, Parveena Bano
ABSTRACT
The present studies were carried out to observe the dissipation pattern of chlorpyrifos on apple in Kashmir Valley. Persistence of chlorpyrifos in apple was studied following two applications rates of chlorpyrifos (Dursban 20 EC) at 200 g a.i. ha(-1) (single dose T 1) and 400 g a.i. ha(-1) (double dose T 2). The average initial deposit of chlorpyrifos was found to be 1.61 and 1.98 μg g(-1) for T 1 and T 2 application rates respectively on apple. The residues dissipated to 0.09 and 0.06 μg g(-1) after 15- and 30-day post treatment with half-life periods of 3.34 and 5.47 days in T 1 and T 2 application rates, respectively. The residues of chlorpyrifos dissipated to below limit of quantification (LOQ) of 0.04 μg g(-1) after 30 day at T 1 application rate. A waiting period of 6 days must be observed for chlorpyrifos on apple at recommended application rate for the safety of consumers. Theoretical maximum residue contribution (TMRC) values were found to be far less than maximum permissible intake (MPI) at 0 day in both the dosages suggesting chlorpyrifos on apple in Kashmir is unlikely to cause health risks.
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ≥98%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium citrate dihydrate, ≥99%, FG
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
Sigma-Aldrich
Sodium citrate dibasic sesquihydrate, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium hydrogencitrate sesquihydrate, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)