Unfortunately, we do not determine the purity of product # 357510. Our specifications only require that we test the overall concentration of the solution.
357510
Phenylethynylmagnesium bromide solution
1.0 M in THF
Synonym(s):
(2-Phenylethynyl)magnesium bromide, (Phenylethynyl)magnesium bromide (6CI), Bromo(2-phenylethynyl)magnesium, Bromo(phenylethynyl)magnesium, Phenylacetylenemagnesium bromide
Select a Size
Select a Size
About This Item
Recommended Products
form
liquid
reaction suitability
reaction type: Grignard Reaction
concentration
1.0 M in THF
density
1.018 g/mL at 25 °C
functional group
phenyl
storage temp.
2-8°C
SMILES string
Br[Mg]C#Cc1ccccc1
InChI
1S/C8H5.BrH.Mg/c1-2-8-6-4-3-5-7-8;;/h3-7H;1H;/q;;+1/p-1
InChI key
KELYMAJHHHGLIW-UHFFFAOYSA-M
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 2
Target Organs
Respiratory system
Supplementary Hazards
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the purity of Phenylethynylmagnesium bromide solution, Product 357510?
1 answer-
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
What type of inert gas is Phenylethynylmagnesium bromide solution, Product 357510, packaged under?
1 answer-
Our packaging department packages product # 357510 under nitrogen.
Helpful?
-
-
What should I do if precipitation is present in my bottle of Phenylethynylmagnesium bromide solution, Product 357510?
1 answer-
We would recommend gently heating the material to allow for the precepitation to redissovle into solution. We have heated the material at 50°C for approximately 2 hours to have the preciptiation redissolve.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service