Skip to Content
Merck
All Photos(1)

Key Documents

346160

Sigma-Aldrich

Ethynylmagnesium chloride solution

0.5 M in THF

Synonym(s):

Chloromagnesium acetylide

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$97.50
800 ML
$316.00

$97.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$97.50
800 ML
$316.00

About This Item

Linear Formula:
CH≡CMgCl
CAS Number:
Molecular Weight:
84.79
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$97.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.921 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Cl[Mg]C#C

InChI

1S/C2H.ClH.Mg/c1-2;;/h1H;1H;/q;;+1/p-1

InChI key

YEWNLGTVYUADOR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ethynylmagnesium chloride solution can be used:
  • To synthesize 1-ethynyldeoxyribose derivatives which can further undergo Sonogashira coupling with various aryl, heteroaryl, and metallocenyl derivatives.[1]
  • In one of the key synthetic steps of substituted naphthostyrils.[2]
  • In the preparation of propargylic ester precursor for the synthesis of 1,3-dienylphosphonates.[3]

recommended

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel and convenient method for the synthesis of substituted naphthostyrils
Liu J, et al.
Tetrahedron Letters, 44(12), 2545-2548 (2003)
Sonogashira reactions of α-and β-1-ethynyl-2-deoxyribosides: synthesis of acetylene-extended C-nucleosides
Bobula T, et al.
Tetrahedron, 66(2), 530-536 (2010)
New and efficient synthesis of 1, 3-dienylphosphonates by Palladium-catalyzed substitution of propargylic esters to diethyl phosphite
Liu X, et al.
Synthetic Communications, 43(19), 2622-2626 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service