Skip to Content
Merck
All Photos(1)

Key Documents

347450

Sigma-Aldrich

Lithium deuteroxide solution

7.5 wt. % in D2O, ≥98 atom % D

Synonym(s):

Lithium deuteroxide (3N in D2O) solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$1,010.00

$1,010.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 G
$1,010.00

About This Item

Linear Formula:
LiOD
CAS Number:
Molecular Weight:
24.95
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.13

$1,010.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥98 atom % D

form

liquid

concentration

7.5 wt. % in D2O

technique(s)

NMR: suitable

refractive index

n20/D 1.363

mass shift

M+1

SMILES string

[Li+].[O-][2H]

InChI

1S/Li.H2O/h;1H2/q+1;/p-1/i/hD

InChI key

WMFOQBRAJBCJND-DYCDLGHISA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service