Skip to Content
Merck
All Photos(2)

Key Documents

316954

Sigma-Aldrich

Ammonium perrhenate

≥99%

Synonym(s):

Metaperrhenic acid ammonium salt, Rhenium standard solution

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$143.00
25 G
$699.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$143.00
25 G
$699.00

About This Item

Linear Formula:
NH4ReO4
CAS Number:
Molecular Weight:
268.24
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder or crystals

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: rhenium

density

3.97 g/mL at 25 °C (lit.)

SMILES string

N.O[Re](=O)(=O)=O

InChI

1S/H3N.H2O.3O.Re/h1H3;1H2;;;;/q;;;;;+1/p-1

InChI key

XPSXHXWHJAIQJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium perrhenate is a white crystalline solid soluble in ethanol and water. It is a weak oxidizer and widely used as a Re precursor in nanomaterial synthesis and Re-based catalyst preparation.

Application

Ammonium perrhenate used:
  • As a rhenium source to prepare Re nanoparticles for catalytic decomposition of ammonium perchlorate.
  • To synthesize ReS2 through a direct sulfidation reaction, for the fabrication of field effect transistors (FETs), digital logic devices, humidity sensors, and rechargeable supercapacitors.
  • As a precursor to prepare the heterogeneous catalysts for organic transformations.
  • An excellent precursor for the fabrication of ReOx/TiO2 recyclable solidcatalyst for deoxydehydration. For at least six runs in a row, the catalyst demonstrated catalyticactivity, selectivity, and stability without deactivation.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perez-Lourido, P. et al.
Inorganic Chemistry, 38, 1511-1511 (1999)
Xuezeng Tian et al.
Nature materials, 19(8), 867-873 (2020-03-11)
The electronic, optical and chemical properties of two-dimensional transition metal dichalcogenides strongly depend on their three-dimensional atomic structure and crystal defects. Using Re-doped MoS2 as a model system, here we present scanning atomic electron tomography as a method to determine
Jeffrey G Linger et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(33), 12013-12018 (2014-08-06)
Lignin is an energy-dense, heterogeneous polymer comprised of phenylpropanoid monomers used by plants for structure, water transport, and defense, and it is the second most abundant biopolymer on Earth after cellulose. In production of fuels and chemicals from biomass, lignin
Yuee Feng et al.
Inorganic chemistry, 48(23), 11058-11066 (2009-11-06)
The detailed syntheses of complexes of the form [Re(O)(X)(RNCH(2)CH(2))(2)N(Me)] (X = Me, Cl, I, R = mesityl, C(6)F(5)), 1-3, incorporating diamidoamine ancillary ligands are described. X-ray crystal structures for the complexes [Re(O)(Me)((C(6)F(5))NCH(2)CH(2))(2)N(Me)], 1a, [Re(O)(I)((C(6)F(5))N CH(2)CH(2))(2)N(Me)], 3a, and [Re(O)(I)((Mes)NCH(2))(2)N(Me)], 3b, are
Rui Cao et al.
Inorganic chemistry, 50(19), 9499-9507 (2011-08-30)
We describe a multidentate tripodal ligand in which three pendant arms carrying di(2-picolyl)amine units are linked to the ortho positions of a tris(o-xylyl) scaffold, providing N(CH(2)-o-C(6)H(4)CH(2)N(CH(2)py)(2))(3) (L). Reaction of L with CuCl(2) in the presence of hexafluorophosphate anion afforded blue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service