Skip to Content
Merck
All Photos(1)

Key Documents

366889

Sigma-Aldrich

Myristic-d27 acid

98 atom % D, 99% (CP)

Synonym(s):

Tetradecanoic-d27 acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,160.00

$1,160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,160.00

About This Item

Linear Formula:
CD3(CD2)12CO2H
CAS Number:
Molecular Weight:
255.54
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$1,160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

mass shift

M+27

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2

InChI key

TUNFSRHWOTWDNC-RZVOLPTOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Titan et al.
Clinica chimica acta; international journal of clinical chemistry, 489, 242-248 (2018-08-29)
Metabolomics can be used to identify novel metabolites related to renal function and that could therefore be used for estimating GFR. We evaluated metabolites replicated and related to eGFR in 3 studies (CKD and general population). Metabolomics was performed by
Silvia M Titan et al.
PloS one, 14(3), e0213764-e0213764 (2019-03-19)
Studies on metabolomics and CKD have primarily addressed CKD incidence defined as a decline on eGFR or appearance of albuminuria in the general population, with very few evaluating hard outcomes. In the present study, we investigated the association between metabolites
Stephan van Vliet et al.
Scientific reports, 11(1), 13828-13828 (2021-07-07)
A new generation of plant-based meat alternatives-formulated to mimic the taste and nutritional composition of red meat-have attracted considerable consumer interest, research attention, and media coverage. This has raised questions of whether plant-based meat alternatives represent proper nutritional replacements to
Solange D Sousa et al.
Molecular reproduction and development, 87(4), 409-418 (2020-03-24)
The present study was conducted to characterize the metabolome of accessory gland fluid (AGF) of locally adapted Morada Nova rams, raised in the Brazilian Northeast. AGF was collected by an artificial vagina from five vasectomized rams. Metabolites were identified by
Teresa Padro et al.
Journal of lipid research, 56(5), 1043-1056 (2015-03-17)
The benefits of dietary phytosterols (PhySs) and long-chain n-3 PUFA (ω3) have been linked to their effects as cholesterol- and triglyceride (TGL)-lowering agents. However, it remains unknown whether these compounds have further metabolic effects on LDL lipid composition. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service