Skip to Content
Merck
All Photos(1)

Key Documents

343609

Sigma-Aldrich

(R)-(+)-Methyl p-tolyl sulfoxide

99%

Synonym(s):

(+)-(R)-Methyl 4-tolyl sulfoxide, (+)-Methyl p-tolyl sulfoxide, (R)-4-Methylphenyl methyl sulfoxide, 1-Methyl-4-[(R)-methylsulfinyl]benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$520.00

$520.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$520.00

About This Item

Linear Formula:
CH3C6H4S(O)CH3
CAS Number:
Molecular Weight:
154.23
Beilstein:
2040677
MDL number:
UNSPSC Code:
12191600
PubChem Substance ID:
NACRES:
NA.22

$520.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

optical activity

[α]20/D +145°, c = 2 in acetone

optical purity

ee: 99% (HPLC)

mp

73-75 °C (lit.)

functional group

sulfoxide

SMILES string

Cc1ccc(cc1)S(C)=O

InChI

1S/C8H10OS/c1-7-3-5-8(6-4-7)10(2)9/h3-6H,1-2H3/t10-/m1/s1

InChI key

FEVALTJSQBFLEU-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-Methyl p-tolyl sulfoxide may be used to prepare (R)-(+)-methyl 3,5-dimethoxy-6-[8-oxo-9-(p-tolylsulfinyl) nonyl] benzoate, an intermediate for (R)-lasiodiplodin synthesis.[1] Its anions undergo addition reaction with nitrones to form optically active a-substituted N-hydroxylamines.[2] (R)-(+)-Methyl p-tolyl sulfoxide also reacts with O-mesitylsulfonylhydroxylamine (MSH) to form (-)-(R)-S-methyl-S-p-tolylsulfoximine.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of orsellinic acid type macrolides: The example of lasiodiplodin.
Solladie G, et al.
Tetrahedron Asymmetry, 1(3), 187-198 (1990)
Chemistry of sulfoxides and related compounds. XLIX. Synthesis of optically active sulfoximines from optically active sulfoxides.
Johnson CR, et al.
The Journal of Organic Chemistry, 39(16), 2458-2459 (1974)
The reaction of nitrones with (R)-(+)-methyl p-tolyl sulfoxide anion; asymmetric synthesis of optically active secondary amines.
Murahashi S-I, et al.
Tetrahedron Letters, 34(16), 2645-2648 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service