Skip to Content
Merck
All Photos(2)

Key Documents

275956

Sigma-Aldrich

Methyl p-tolyl sulfide

99%

Synonym(s):

4-(Methylthio)toluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$91.20
25 G
$350.00

$91.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$91.20
25 G
$350.00

About This Item

Linear Formula:
CH3C6H4SCH3
CAS Number:
Molecular Weight:
138.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$91.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.573 (lit.)

bp

52-54 °C/1 mmHg (lit.)

density

1.027 g/mL at 25 °C (lit.)

functional group

thioether

SMILES string

CSc1ccc(C)cc1

InChI

1S/C8H10S/c1-7-3-5-8(9-2)6-4-7/h3-6H,1-2H3

InChI key

VHILIAIEEYLJNA-UHFFFAOYSA-N

General description

Aerobic oxidation of methyl p-tolyl sulfide catalyzed by a heteroscorpionate Ru(II)-aqua complex has been reported[1]. Chloroperoxidase catalyzed oxidation of methyl p-tolyl sulfide has been reported[2].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chloroperoxidase and hydrogen peroxide: An efficient system for enzymatic enantioselective sulfoxidations
Colonna S, et al.
Tetrahedron Asymmetry, 3(1), 95-106 (1992)
My Hang V Huynh et al.
Journal of the American Chemical Society, 125(2), 308-309 (2003-01-09)
fac-[RuII(Cl)(dpp)(L3)]+ (L3 = tris(pyrid-2-yl)methoxymethane (tpmm) = [1A]+ and tris(pyrid-2-yl)pentoxymethane (tppm) = [1B]+ and dpp = di(pyrazol-1-yl)propane) rapidly undergo ligand substitution with water to form fac-[RuII(H2O)(dpp)(L3)]2+ (L3 = tpmm = [2A]2+ and tppm = [2B]2+). In the structure of [2A]2+, the
Vasiliy Yu Evtushok et al.
Frontiers in chemistry, 7, 858-858 (2020-01-11)
In this work, we elaborated heterogeneous catalysts on the basis of the Venturello complex [PO4{WO(O2)2}4]3- (PW4) and nitrogen-free or nitrogen-doped carbon nanotubes (CNTs or N-CNTs) for epoxidation of alkenes and sulfoxidation of thioethers with aqueous hydrogen peroxide. Catalysts PW4/CNTs and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service