Skip to Content
Merck
All Photos(1)

Key Documents

339997

Sigma-Aldrich

(S)-(−)-Methyl p-tolyl sulfoxide

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$711.00

$711.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$711.00

About This Item

Linear Formula:
CH3C6H4S(O)CH3
CAS Number:
Molecular Weight:
154.23
Beilstein:
2324696
MDL number:
UNSPSC Code:
12191600
PubChem Substance ID:
NACRES:
NA.22

$711.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

optical activity

[α]20/D −145°, c = 2 in acetone

optical purity

ee: 99% (HPLC)

mp

75-77 °C (lit.)

SMILES string

Cc1ccc(cc1)S(C)=O

InChI

1S/C8H10OS/c1-7-3-5-8(6-4-7)10(2)9/h3-6H,1-2H3/t10-/m0/s1

InChI key

FEVALTJSQBFLEU-JTQLQIEISA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(-)-Methyl p-tolyl sulfoxide can be used as a nucleophilic reagent to synthesize:
  • Optically active β-disulfoxides by reacting with arenesulfinic esters via formation of α-sulfinylcarbanion.[1]
  • α-substituted N-hydroxylamines by treating with nitrones via preparation of (S)-(-)-methyl p-tolyl sulfoxide anion.[2]
  • 2-O-benzyl-3,4-O-isopropylidene-L-erythrose by one-carbon homologation of 2,3-O-isopropylidene-L-glyceraldehyde.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

β -Disulfoxides. II. The Preparation of Some Optically Active β -Disulfoxides
Kunieda N, et al.
Bulletin of the Chemical Society of Japan, 49(1), 256-259 (1976)
A highly stereoselective synthesis of d-erythrose derivatives by one-carbon homologation of 2, 3-O-isopropylidene-d-glyceraldehyde with (R)-methyl p-tolyl sulfoxide.
Arroyo-Gomez Y, et al.
Tetrahedron Asymmetry, 11(3), 789-796 (2000)
The reaction of nitrones with (R)-(+)-methyl p-tolyl sulfoxide anion; asymmetric synthesis of optically active secondary amines.
Murahashi S-I, et al.
Tetrahedron Letters, 34(16), 2645-2648 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service