Skip to Content
Merck
All Photos(1)

Key Documents

309753

Sigma-Aldrich

(R)-(−)-3-Hydroxytetrahydrofuran

98%

Synonym(s):

(R)-(-)-Tetrahydro-3-furanol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$57.80

$57.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$57.80

About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$57.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

optical activity

[α]20/D −18°, c = 2.4 in methanol

refractive index

n20/D 1.45 (lit.)

bp

181 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

O[C@@H]1CCOC1

InChI

1S/C4H8O2/c5-4-1-2-6-3-4/h4-5H,1-3H2/t4-/m1/s1

InChI key

XDPCNPCKDGQBAN-SCSAIBSYSA-N

Application

(R)-(-)-3-Hydroxytetrahydrofuran may be used as building block in the synthesis of potent HIV protease inhibitors.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diethyl zinc catalyzed diastereoselective addition of ketenes to (S)-(+)-3-hydroxytetrahydrofuran.
Tandon VK.
Tetrahedron Letters, 42(34), 5985-5987 (2001)
Angle-Resolved Photoelectron Spectroscopy of Randomly Oriented 3-Hydroxytetrahydrofuran Enantiomers.
Giardini A, et al.
ChemPhysChem, 6(6), 1164-1168 (2005)
Matt Hrapchak et al.
Journal of labelled compounds & radiopharmaceuticals, 57(12), 687-694 (2014-10-22)
Empagliflozin, (2S,3R,4R,5S,6R)-2-[4-chloro-3-[[4-[(3S)-oxolan-3-yl]oxyphenyl]methyl]phenyl]-6-(hydroxymethyl)oxane-3,4,5-triol was recently approved by the FDA for the treatment of chronic type 2 diabetes mellitus. Herein, we report the synthesis of carbon-13 and carbon-14 labeled empagliflozin. Carbon-13 labeled empagliflozin was prepared in five steps and in 34% overall

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service