Skip to Content
Merck
All Photos(2)

Key Documents

337927

Sigma-Aldrich

Ytterbium(III) chloride hexahydrate

99.9% trace metals basis

Synonym(s):

Ytterbium trichloride hydrate, Ytterbium(III) chloride·6hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$80.50
50 G
$320.00

$80.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$80.50
50 G
$320.00

About This Item

Linear Formula:
YbCl3 · 6H2O
CAS Number:
Molecular Weight:
387.49
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystals and lumps

$80.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Assay

99.9% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: ytterbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

mp

180 °C (dec.) (lit.)

density

2.58 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Yb](Cl)Cl

InChI

1S/3ClH.6H2O.Yb/h3*1H;6*1H2;/q;;;;;;;;;+3/p-3

InChI key

LEYFXTUKPKKWMP-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Ytterbium(III) chloride hexahydrate can be used in applications such as: rare earth doping of yttrium oxide (Y2O3) nanoparticles for the development of fluorescent materials[1], doping of yttrium fluoride (YF3) nanoparticles for potential usage in 3D displays, photovoltaics, and drug delivery[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lanthanide-doped ultrasmall yttrium fluoride nanoparticles with enhanced multicolor upconversion photoluminescence
Chen G, et al.
Journal of Materials Chemistry, 22(38), 20190-20196 (2012)
Rare-earth-doped and codoped Y 2O 3 nanomaterials as potential bioimaging probes.
Das GK and Tan TTY
The Journal of Physical Chemistry C, 112(30), 11211-11217 (2008)
Yasuchika Hasegawa et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(53), 12308-12315 (2019-07-23)
Oxygen-sensitive and near-infrared (NIR) luminescent YbIII coordination polymers incorporating ligands based on pyrene derivatives were synthesized: YbIII -TBAPy and YbIII -TIAPy (TBAPy: 1,3,6,8-tetrakis(p-benzoate)pyrene; TIAPy: 1,3,6,8-tetrakis(3,5-isophthalic acid)pyrene). The coordination structures of these materials have been characterized by means of electrospray ionization
Yingying Li et al.
Frontiers in chemistry, 8, 832-832 (2020-11-12)
The rare-earth nanocrystals containing Er3+ emitters offer very promising tools for imaging applications, as they can not only exhibit up-conversion luminescence but also down-conversion luminescence in the second near-infrared window (NIR II). Doping non-lanthanide cations into host matrix was demonstrated
Uliana Kostiv et al.
Beilstein journal of nanotechnology, 6, 2290-2299 (2016-01-07)
NaYF4:Yb(3+)/Er(3+) nanoparticles were synthesized by thermal decomposition of lanthanide trifluoroacetates using oleylamine (OM) as both solvent and surface binding ligand. The effect of reaction temperature and time on the properties of the particles was investigated. The nanoparticles were characterized by

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service