336920
Isopropyldiphenylphosphine
97%
Synonym(s):
NSC 244305
Select a Size
About This Item
Recommended Products
Assay
97%
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
bp
165 °C/11 mmHg (lit.)
mp
40-43 °C (lit.)
functional group
phosphine
SMILES string
CC(C)P(c1ccccc1)c2ccccc2
InChI
1S/C15H17P/c1-13(2)16(14-9-5-3-6-10-14)15-11-7-4-8-12-15/h3-13H,1-2H3
InChI key
LLZAIAIZAVMQIG-UHFFFAOYSA-N
Application
- Nickel/Lewis acid-catalyzed aryl- and alkenylcyanation of unsaturated bonds
- Nickel/Lewis Acid-Catalyzed Cyanoesterification and Cyanocarbamoylation of Alkynes
- Methoxycarbonylation reactions (palladium complex)
- Decarboxylative cross-coupling in the presence of palladium
- Asymmetric hydrogenation of quinolines catalyzed by iridium complexes of BINOL-derived diphosphonites
- Hydroesterification of vinylarenes in complex with palladium
- Ligand precatalyst for room-temperature Heck coupling reactions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service