Skip to Content
Merck
All Photos(1)

Key Documents

591688

Sigma-Aldrich

tert-Butyldiphenylphosphine

97%

Synonym(s):

NSC 244302

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2PC(CH3)3
CAS Number:
Molecular Weight:
242.30
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

bp

144-146 °C/2 mmHg (lit.)

mp

52-57 °C (lit.)

functional group

phosphine

SMILES string

CC(C)(C)P(c1ccccc1)c2ccccc2

InChI

1S/C16H19P/c1-16(2,3)17(14-10-6-4-7-11-14)15-12-8-5-9-13-15/h4-13H,1-3H3

InChI key

QZUPHAGRBBOLTB-UHFFFAOYSA-N

Application

Catalyst for:
  • Nickel/Lewis acid-catalyzed carbocyanation of alkynes with acetonitrile or substituted acetonitriles
  • Palladium catalyzed hydrocarboxylation of acetylene with carbon monoxide to acrylic acid under mild conditions
  • Palladium to form a catalyst for methoxycarbonylation reactions
  • Ru-catalyzed transfer hydrogenation in reductive coupling of disubstituted allenes with aldehydes
  • Stereoselective silylation by dehydrogenative Si-O coupling with Si-stereogenic silanes

  • Phosphine ligand in nickel-catalyzed carbocyanation of alkynes
  • Monodentate P-Donor Ligands (LKB-P)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Giovanni Ricci et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-27)
Two novel cobalt diphenylphosphine complexes were synthesized by reacting cobalt(II) chloride with tert-butyl(diphenyl)phosphine (P t

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service