D39207
trans-1,4-Dibromo-2-butene
99%
Synonym(s):
(2E)-1,4-Dibromo-2-butene, (2E)-1,4-Dibromobut-2-ene, 1,4-Dibromo-2-(E)-butene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
BrCH2CH=CHCH2Br
CAS Number:
Molecular Weight:
213.90
Beilstein:
1719696
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
crystals
bp
205 °C (lit.)
mp
48-51 °C (lit.)
storage temp.
2-8°C
SMILES string
BrC\C=C\CBr
InChI
1S/C4H6Br2/c5-3-1-2-4-6/h1-2H,3-4H2/b2-1+
InChI key
RMXLHIUHKIVPAB-OWOJBTEDSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Skin Corr. 1B
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Xiuzhong Zhu et al.
Polymers, 11(4) (2019-04-10)
A series of side-chain functionalized polytetrahydrofuran (PTHF) derivatives were synthesized via the blue-light photocatalytic thiol-ene "click" reaction. Firstly, unsaturated polytetrahydrofuran (UPTHF) as a new unsaturated polyether was synthesized via condensation polymerization of cis-2-butene-1,4-diol and trans-1,4-dibromo-2-butene using potassium hydroxide (KOH) as
Kwei-Yu Liu et al.
Polymers, 11(5) (2019-05-07)
Localized gene delivery still remains as a challenging therapeutic method due to the multiple hurdles to overcome. One of the significant factors is a development of a matrix to carry and safely deliver genes at the local site in a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service