Skip to Content
Merck
All Photos(1)

Key Documents

333441

Sigma-Aldrich

Methyl 4-(trifluoromethyl)benzoate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$91.20

$91.20


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$91.20

About This Item

Linear Formula:
CF3C6H4CO2CH3
CAS Number:
Molecular Weight:
204.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$91.20


Estimated to ship onMay 30, 2025


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.451 (lit.)

bp

94-95 °C/21 mmHg (lit.)

mp

13-14 °C (lit.)

density

1.268 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

COC(=O)c1ccc(cc1)C(F)(F)F

InChI

1S/C9H7F3O2/c1-14-8(13)6-2-4-7(5-3-6)9(10,11)12/h2-5H,1H3

InChI key

VAZWXPJOOFSNLB-UHFFFAOYSA-N

General description

Methyl 4-(trifluoromethyl)benzoate is an ester. The reduction of methyl (4-trifluoromethyl)benzoate catalyzed by MoO2Cl2 was reported.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silane/ MoO2Cl2 as an efficient system for the reduction of esters.
Fernandes AC and Rom?o CC.
J. Mol. Catal. A: Chem., 253(1), 96-98 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service