Skip to Content
Merck
All Photos(1)

Key Documents

483877

Sigma-Aldrich

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate

97%

Synonym(s):

S-(Trifluoromethyl)dibenzothiophenium triflate, S-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$166.00

$166.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$166.00

About This Item

Empirical Formula (Hill Notation):
C14H8F6O3S2
CAS Number:
Molecular Weight:
402.33
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$166.00


Estimated to ship onMarch 23, 2025


Request a Bulk Order

Quality Level

Assay

97%

mp

150 °C (lit.)

functional group

fluoro
triflate

SMILES string

[O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1c2ccccc2-c3ccccc13

InChI

1S/C13H8F3S.CHF3O3S/c14-13(15,16)17-11-7-3-1-5-9(11)10-6-2-4-8-12(10)17;2-1(3,4)8(5,6)7/h1-8H;(H,5,6,7)/q+1;/p-1

InChI key

QXXHXTRTGZBOGD-UHFFFAOYSA-M

Application

  • Pd(II)-catalyzed trifluoromethylation[1]
  • Electrophilic trifluoromethylation in ionic liquids[2]
  • Used in the stereoselective preparation of trifluoromethylalkynes by trifluoromethylation of terminal alkynes using Umemoto′s reagent and a copper catalyst[3][4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ionic liquids as new media for electrophilic trifluoromethylation reactions
Pegot, B.; et al.
Journal of Fluorine Chemistry, 134, 156-159 (2012)
Copper-catalyzed trifluoromethylation of terminal alkynes using Umemoto's reagent
Luo, D.-F.; et al.
Tetrahedron Letters, 53, 2769-2772 (2012)
Jun Xu et al.
Journal of the American Chemical Society, 133(39), 15300-15303 (2011-09-15)
An unprecedented type of reaction for Cu-catalyzed trifluoromethylation of terminal alkenes is reported. This reaction represents a rare instance of catalytic trifluoromethylation through C(sp(3))-H activation. It also provides a mechanistically unique example of Cu-catalyzed allylic C-H activation/functionalization. Both experimental and
Xing-Guo Zhang et al.
Journal of the American Chemical Society, 134(29), 11948-11951 (2012-07-12)
A Pd(II)-catalyzed trifluoromethylation of ortho C-H bonds with an array of N-arylbenzamides derived from benzoic acids is reported. N-Methylformamide has been identified as a crucial promoter of C-CF(3) bond formation from the Pd center. X-ray characterization of the C-H insertion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service