Skip to Content
Merck
All Photos(2)

Documents

332364

Sigma-Aldrich

Allyltriphenylstannane

97%

Synonym(s):

2-Propenyl-triphenylstannane, Allyltriphenyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2Sn(C6H5)3
CAS Number:
Molecular Weight:
391.09
Beilstein:
3612762
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

71-74 °C (lit.)

SMILES string

C=CC[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.C3H5.Sn/c3*1-2-4-6-5-3-1;1-3-2;/h3*1-5H;3H,1-2H2;

InChI key

NDUYAGLANMHJHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyltriphenylstannane is an organotin compound, which is generally used as allylating and radical chain transfer reagent. It is also used as a source of allyl radicals.

Application

Allyltriphenylstannane can be used:
  • As a reagent in the C-H phenylation of azoles catalyzed by palladium.
  • As an allylating reagent in the alkylidene Meldrum′s acids allylation catalyzed by Sc(OTf)3.
  • As a reagent in the total synthesis of an antifungal molecule (+)-ambruticin S.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of (+)-ambruticin S: probing the pharmacophoric subunit
Hanessian S, et al.
The Journal of Organic Chemistry, 75(16), 5601-5618 (2010)
Palladium-Catalyzed Direct Arylations of Azoles with Aryl Silicon and Tin Reagents
Han W, et al.
Chemistry?A European Journal , 17(25), 6904-6908 (2011)
Allyltriphenylstannane
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Sc (OTf) 3-Catalyzed Conjugate Allylation of Alkylidene Meldrum′s Acids
Dumas AM and Fillion E
Organic Letters, 11(9), 1919-1922 (2009)
Journal of the American Chemical Society, 103, 1969-1969 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service