Skip to Content
Merck
All Photos(2)

Key Documents

271411

Sigma-Aldrich

Allyltributylstannane

97%

Synonym(s):

Allyltri-n-butyltin, Allyltributyltin, Tributyl-2-propenylstannane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$81.20
25 G
$176.00
100 G
$375.00

$81.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$81.20
25 G
$176.00
100 G
$375.00

About This Item

Linear Formula:
CH2=CHCH2Sn(CH2CH2CH2CH3)3
CAS Number:
Molecular Weight:
331.12
Beilstein:
3588340
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$81.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.486 (lit.)

bp

88-92 °C/0.2 mmHg (lit.)

density

1.068 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)CC=C

InChI

1S/3C4H9.C3H5.Sn/c3*1-3-4-2;1-3-2;/h3*1,3-4H2,2H3;3H,1-2H2;

InChI key

YLGRTLMDMVAFNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allylation reagent for aldehydes catalyzed by chiral Lewis acids[1][2] and ytterbium(III) trifluoromethanesulfonate (cat. no. 430595).[3] Undergoes tetrakis(triphenylphosphine)palladium(0) (cat. no. 216666) catalyzed coupling with iodoquinones[4] and allyl acetates.[5][6]
Efficient allylation of aldehydes leading to homoallylic alcohols with trichloro-1,3,5-triazene.[7]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

217.4 °F - closed cup

Flash Point(C)

103 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Journal of the American Chemical Society, 115, 8467-8467 (1993)
Tetrahedron Letters, 34, 7827-7827 (1993)
Aldrichimica Acta, 20, 45-45 (1987)
Tetrahedron Letters, 47, 9103-9103 (2006)
Journal of the American Chemical Society, 35, 4639-4639 (1994)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service