Skip to Content
Merck
All Photos(3)

Key Documents

374601

Sigma-Aldrich

Tributylphenylstannane

97%

Synonym(s):

Phenyltributylstannane, Phenyltributyltin, Tributylphenyltin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$80.80
10 G
$242.00

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$80.80
10 G
$242.00

About This Item

Linear Formula:
C6H5Sn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
367.16
Beilstein:
3610571
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.516 (lit.)

bp

125-128 °C/0.14 mmHg (lit.)

density

1.125 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)c1ccccc1

InChI

1S/C6H5.3C4H9.Sn/c1-2-4-6-5-3-1;3*1-3-4-2;/h1-5H;3*1,3-4H2,2H3;

InChI key

SYUVAXDZVWPKSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service