Skip to Content
Merck
All Photos(1)

Key Documents

331910

Sigma-Aldrich

2-Azabicyclo[2.2.1]hept-5-en-3-one

98%

Synonym(s):

(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 4-Amino-2-cyclopentene-1-carboxylic acid lactam

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$49.80

$49.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$49.80

About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
508342
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$49.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Assay

98%

bp

102-106 °C/0.25 mmHg (lit.)

mp

54-58 °C (lit.)

SMILES string

O=C1N[C@H]2C[C@@H]1C=C2

InChI

1S/C6H7NO/c8-6-4-1-2-5(3-4)7-6/h1-2,4-5H,3H2,(H,7,8)/t4-,5+/m0/s1

InChI key

DDUFYKNOXPZZIW-CRCLSJGQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Azabicyclo[2.2.1]hept-5-en-3-one is also referred as vince lactam.[1] It is a versatile intermediate in the synthesis of carbocyclic nucleosides.[2] 2-Azabicyclo[2.2.1]hept-5-en-3-one and its monohydrated complex was investigated in a supersonic jet by Fourier transform microwave spectroscopy.[1]

Application

2-Azabicyclo[2.2.1]hept-5-en-3-one was used in:
  • preparation of (3aS,4R,6R,6aR)-6-((methoxy-methoxy)methyl)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-amine, a precursor of analog of bredinin[3]
  • synthesis of therapeutic drugs[1]
  • chemoenzymatic synthesis of (−)-carbovir[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Écija et al.
The journal of physical chemistry. A, 116(41), 10099-10106 (2012-09-19)
2-Azabicyclo[2.2.1]hept-5-en-3-one (ABH or Vince lactam) and its monohydrated complex (ABH···H(2)O) have been observed in a supersonic jet by Fourier transform microwave spectroscopy. ABH is broadly used in the synthesis of therapeutic drugs, whereas the ABH···H(2)O system offers a simple model
Vasu Nair et al.
Molecules (Basel, Switzerland), 18(9), 11576-11585 (2013-09-21)
The natural nucleoside antibiotic, bredinin, exhibits antiviral and other biological activities. While various nucleosides related to bredinin have been synthesized, its carbocyclic analog has remained unknown. Synthesis of this heretofore unknown analog of bredinin is described. The key precursor, (3aS,4R,6R,6aR)-6-((methoxy-methoxy)methyl)-2,2-dimethyltetrahydro-3aH-cyclopenta[d][1,3]dioxol-4-amine
Chemoenzymatic synthesis of (-)-carbovir utilizing a whole cell catalysed resolution of 2-azabicyclo [2.2. 1] hept-5-en-3-one.
Steven, J. C.
Journal of the Chemical Society. Chemical Communications, 16, 1120-1121 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service