Skip to Content
Merck
All Photos(1)

Key Documents

155268

Sigma-Aldrich

(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid

99%

Synonym(s):

(+)-MTPA, Mosher’s acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$113.00
1 G
$379.00
5 G
$1,720.00

$113.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$113.00
1 G
$379.00
5 G
$1,720.00

About This Item

Linear Formula:
C6H5C(OCH3)(CF3)CO2H
CAS Number:
Molecular Weight:
234.17
Beilstein:
3591560
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

$113.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

optical activity

[α]20/D +72°, c = 1.6 in methanol

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.473 (lit.)

bp

105-107 °C/1 mmHg (lit.)

mp

46-49 °C (lit.)

density

1.344 g/mL at 25 °C (lit.)

functional group

carboxylic acid
ether
fluoro
phenyl

storage temp.

2-8°C

SMILES string

CO[C@@](C(O)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H9F3O3/c1-16-9(8(14)15,10(11,12)13)7-5-3-2-4-6-7/h2-6H,1H3,(H,14,15)/t9-/m1/s1

InChI key

JJYKJUXBWFATTE-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(+)-α-Methoxy-α-trifluoromethylphenylacetic acid is commonly used as a derivatizing agent in Mosher ester analysis, an NMR-based method for determining the absolute configuration of the chiral carbon center in a secondary alcohol.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

doi:10.1038/nprot.2007.354

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tetrahedron Asymmetry, 18, 975-975 (2007)
Lidia De Luca et al.
The Journal of organic chemistry, 72(10), 3955-3957 (2007-04-11)
An effective route to chiral optically active 2-substituted benzofurans directly from carboxylic acids is reported. This procedure, which allows the preparation of alpha-alkyl-2-benzofuranmethanamines from N-protected alpha-amino acids without sensible racemization phenomena, proceeds in good yields under mild conditions with the
Mosher ester analysis for the determination of absolute configuration of stereogenic (chiral) carbinol carbons.
Hoye TR, et al.
Nature Protocols, 2(10), 2451-2458 (2007)
J Martín Torres-Valencia et al.
Phytochemical analysis : PCA, 13(6), 329-332 (2002-12-24)
A method to determine the absolute configuration of 2,3-epoxy-2-methylbutanoate ester residues in natural products is presented, based on (i) the reduction of the ester function to yield a 2-methyl-1,2-butanediol, (ii) esterification of the obtained primary alcohol with either (R)-(+)- or
Giancarlo Cravotto et al.
Chirality, 16(8), 526-533 (2004-08-04)
Cyclodextrin (CD) derivatives are important selectors for analytical chiral recognition. Their enantioselectivities and chemical properties depend on ring size and on nature, number and location of substituents. This paper describes the synthesis of 6-O-TBDMS-2,3-O-methyl beta-cyclodextrins bearing in position 2 either

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service