Skip to Content
Merck
All Photos(1)

Key Documents

441279

Sigma-Aldrich

(1R)-(−)-2-Azabicyclo[2.2.1]hept-5-en-3-one

≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$44.50
5 G
$193.00

$44.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$44.50
5 G
$193.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
4230721
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$44.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

solid

optical activity

[α]20/D −565°, c = 1 in chloroform

optical purity

ee: 99% (HPLC)

mp

94-97 °C (lit.)

SMILES string

O=C1N[C@@H]2C[C@H]1C=C2

InChI

1S/C6H7NO/c8-6-4-1-2-5(3-4)7-6/h1-2,4-5H,3H2,(H,7,8)/t4-,5+/m1/s1

InChI key

DDUFYKNOXPZZIW-UHNVWZDZSA-N

General description

(1R)-(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one is a bicyclic γ-lactam.[1]

Application

(1R)-(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one can be used as a precursor to prepare:
  • Amino-peramivir, a potent neuraminidase inhibitor.[2]
  • Five membered analogs of 4-amino-5-halopentanoic acids as potential GABA aminotransferase (GABA-AT) inactivators.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new class of conformationally rigid analogues of 4-amino-5-halopentanoic acids, potent inactivators of ?-aminobutyric acid aminotransferase.
Qiu J and Silverman RB.
Journal of Medicinal Chemistry, 43(4), 706-720 (2000)
The de-guanidinylated derivative of peramivir remains a potent inhibitor of influenza neuraminidase
Bromba CM, et al.
Bioorganic & medicinal chemistry letters, 21(23), 7137-7141 (2011)
2-Azabicyclo [2.2. 1] hept-5-en-3-one: chemical profile of a versatile synthetic building block and its impact on the development of therapeutics.
Singh R and Vince R.
Chemical Reviews, 112(8), 4642-4686 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service