Skip to Content
Merck
All Photos(1)

Key Documents

299936

Sigma-Aldrich

2-Amino-7-bromofluorene

90%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$74.80

$74.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$74.80

About This Item

Empirical Formula (Hill Notation):
C13H10BrN
CAS Number:
Molecular Weight:
260.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$74.80

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

90%

form

powder

mp

148-149 °C (lit.)

functional group

bromo

SMILES string

Nc1ccc-2c(Cc3cc(Br)ccc-23)c1

InChI

1S/C13H10BrN/c14-10-1-3-12-8(6-10)5-9-7-11(15)2-4-13(9)12/h1-4,6-7H,5,15H2

InChI key

RJWYTISHBMNMOZ-UHFFFAOYSA-N

General description

Spectral characteristics of inclusion complexation of 2-amino-7-bromofluorene in aqueous β-cyclodextrin solution were evaluated.[1] Fluorescence quenching of 2-amino-7-bromofluorene by chloromethanes (CH2Cl2, CHCl3, CCl4) in various solvents was studied.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescence quenching of 2-amino-7-bromofluorene by chloromethanes-static and dynamic model for CCl4 quenching in polar solvents.
Radha N and Swaminathan M.
J. Chem. Res. (M), 2001(12), 503-504 (2001)
I V Muthu Vijayan Enoch et al.
Journal of fluorescence, 14(6), 751-756 (2005-01-15)
Spectral characteristics of 2-amino-7-bromofluorene (2ABF) have been studied in aqueous beta-cyclodextrin (beta-CDx) solution. Enhancement in the fluorescence intensity of the neutral from of 2ABF was observed due to the formation of 1:1 complex with beta-CDx. The formation of this complex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service