Merck
  • Divergent reaction pathways of homologous and isosteric propargyl amides in sequential Ru/Pd-catalyzed annulations for the synthesis of heterocycles.
Divergent reaction pathways of homologous and isosteric propargyl amides in sequential Ru/Pd-catalyzed annulations for the synthesis of heterocycles.
The Journal of organic chemistry (2013-03-26)
Sandeep N Raikar, Helena C Malinakova
ABSTRACT
Cu-catalyzed three-component coupling of imines with benzoyl chloride and terminal arylalkynes followed by enyne ring-closing metathesis (RCM) and Heck cyclization afforded medicinally relevant benzoindolines, cyclopropane-fused indenopyridines, pyrroloquinolines, or 1,7-tetrahydrophenanthrolines via divergent cyclization pathways. Unexpectedly, the Pd-catalyzed cyclization of heterocyclic dienes proceeded via regiodivergent 5-exo or 6-endo pathways depending on the ring size (n = 1, 2) or the presence of isosteric groups (CH vs N). A one-pot protocol for the enyne-RCM/Heck annulation featuring a sequential addition of the Ru and Pd catalysts was developed maximizing the synthetic efficiency.
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
N-Methyl-N-propargylbenzylamine, 97%
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Pargyline hydrochloride
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%