1.00990
Ethanol
dried (max. 0.01% H2O)
Synonym(s):
Ethyl alcohol
About This Item
59 hPa ( 20 °C)
Recommended Products
grade
anhydrous
vapor density
1.59 (vs air)
vapor pressure
44.6 mmHg ( 20 °C)
59 hPa ( 20 °C)
Assay
≥99.9% (GC)
form
liquid
autoignition temp.
683 °F
potency
6200 mg/kg LD50, oral (Rat)
expl. lim.
19 %, 60 °F
impurities
≤0.0002 meq/g Alkalinity
≤0.0005 meq/g Acidity
≤0.01% 2-Propanol (GC)
≤0.01% Acetone (GC)
≤0.01% Water
≤0.05% Methanol (GC)
evapn. residue
≤10 mg/L
color
APHA: ≤10
refractive index
n20/D 1.3600 (lit.)
pH
7.0 (20 °C, 10 g/L in H2O)
bp
78 °C (lit.)
mp
−114 °C (lit.)
transition temp
flash point 12 °C
application(s)
pharmaceutical
format
neat
storage temp.
2-30°C
SMILES string
CCO
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3
InChI key
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Analysis Note
Identity (IR): conforms
Colour: ≤ 10 Hazen
Acidity: ≤ 0.0005 meq/g
Alkalinity: ≤ 0.0002 meq/g
Acetone (GC): ≤ 0.01 %
Methanol (GC): ≤ 0.05 %
2-Propanol (GC): ≤ 0.01 %
Evaporation residue: ≤ 10 mg/l
Water: ≤ 0.01 %
Footnote
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
55.4 °F
Flash Point(C)
13 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service