Skip to Content
Merck
All Photos(2)

Key Documents

317365

Sigma-Aldrich

Tetramethylammonium triacetoxyborohydride

95%

Synonym(s):

triacetoxyboranuide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$269.00
50 G
$765.00

$269.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$269.00
50 G
$765.00

About This Item

Linear Formula:
(CH3)4N(CH3CO2)3BH
CAS Number:
Molecular Weight:
263.10
Beilstein:
8238791
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$269.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

95%

form

solid

reaction suitability

reagent type: reductant

mp

93-98 °C (lit.)

functional group

amine

SMILES string

C[N+](C)(C)C.CC(=O)O[BH-](OC(C)=O)OC(C)=O

InChI

1S/C6H10BO6.C4H12N/c1-4(8)11-7(12-5(2)9)13-6(3)10;1-5(2,3)4/h7H,1-3H3;1-4H3/q-1;+1

InChI key

LCFZZOGKVOTFPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selective reducing agent.[1][2][3]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 44, 7239-7243 (2003)
Chemtracts, 5, 188-192 (1992)
Ning Shangguan et al.
Organic letters, 9(6), 1093-1096 (2007-02-27)
A formal synthesis of psymberin (irciniastatin A) is presented. Notable features of the synthesis include the chemo-, regio-, and stereoselective oxidation of a 1,3-disubstituted allene, a configuration-dependent spirodiepoxide opening, the efficient syntheses of functionalized trans-2,6-disubstituted pyrans, and the union of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service