Skip to Content
Merck
Tetramethylammonium triacetoxyborohydride 95%

Tetramethylammonium triacetoxyborohydride

Tetramethylammonium triacetoxyborohydride 95%
Linear Formula:
(CH3)4N(CH3CO2)3BH
CAS Number:
Molecular Weight:
263.10

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing