Skip to Content
Merck
All Photos(1)

Key Documents

310808

Sigma-Aldrich

D-Glucose-6-13C

99 atom % 13C

Synonym(s):

Labeled Glucose, Dextrose-6-13C

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,010.00

$1,010.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more informationRequest more information

Select a Size

Change View
100 MG
$1,010.00

About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
Beilstein:
5810734
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$1,010.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more informationRequest more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+1

SMILES string

O[13CH2][C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1/i2+1

InChI key

GZCGUPFRVQAUEE-HYISWWFJSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service