Skip to Content
Merck
All Photos(1)

Key Documents

453196

Sigma-Aldrich

D-Glucose-1,6-13C2

≥99 atom % 13C, ≥99% (CP)

Synonym(s):

Labeled Glucose, Dextrose-1,6-13C2

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$1,440.00
250 MG
$3,220.00

$1,440.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
$1,440.00
250 MG
$3,220.00

About This Item

Empirical Formula (Hill Notation):
13C2C4H12O6
CAS Number:
Molecular Weight:
182.14
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.12

$1,440.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

≥99 atom % 13C

Quality Level

Assay

≥99% (CP)

form

powder

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C

mass shift

M+2

storage temp.

room temp

SMILES string

O[13CH2][C@@H]1O[13CH2][C@@H](O)[C@H](O)[C@H]1O.O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6+/m0/s1/i1+1,2+1

InChI key

GZCGUPFRVQAUEE-OFAMVBKFSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tigran V Yuzbashev et al.
Biotechnology and bioengineering, 113(11), 2425-2432 (2016-05-18)
Bio-based succinic acid production can redirect industrial chemistry processes from using limited hydrocarbons to renewable carbohydrates. A fermentation process that does not require pH-titrating agents will be advantageous to the industry. Previously, a Yarrowia lipolytica strain that was defective for
Xuemei Tong et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(51), 21660-21665 (2009-12-10)
Tumor cells are metabolically reprogrammed to fuel cell proliferation. Most transformed cells take up high levels of glucose and produce ATP through aerobic glycolysis. In cells exhibiting aerobic glycolysis, a significant fraction of glucose carbon is also directed into de
Ralph J DeBerardinis et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(49), 19345-19350 (2007-11-23)
Tumor cell proliferation requires rapid synthesis of macromolecules including lipids, proteins, and nucleotides. Many tumor cells exhibit rapid glucose consumption, with most of the glucose-derived carbon being secreted as lactate despite abundant oxygen availability (the Warburg effect). Here, we used
Barrett L Updegraff et al.
Cell reports, 25(8), 2223-2233 (2018-11-22)
Pathways underlying metabolic reprogramming in cancer remain incompletely understood. We identify the transmembrane serine protease TMPRSS11B as a gene that promotes transformation of immortalized human bronchial epithelial cells (HBECs). TMPRSS11B is upregulated in human lung squamous cell carcinomas (LSCCs), and high

Articles

Isotopic Labeling for NMR Spectroscopy of Biological Solids;

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service