Skip to Content
Merck
All Photos(1)

Key Documents

310794

Sigma-Aldrich

D-Glucose-2-13C

99 atom % 13C

Synonym(s):

Labeled Glucose, Dextrose-2-13C

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$817.00
1 G
$2,330.00

$817.00


Estimated to ship onMay 28, 2025


Request more information

Select a Size

Change View
250 MG
$817.00
1 G
$2,330.00

About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$817.00


Estimated to ship onMay 28, 2025


Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

powder

optical activity

[α]25/D +52.0, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

M+1

SMILES string

OC[C@H]1O[C@H](O)[13C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i5+1

InChI key

WQZGKKKJIJFFOK-UKLRSMCWSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Isotopic Labeling for NMR Spectroscopy of Biological Solids;

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service