Skip to Content
Merck
All Photos(1)

Key Documents

605131

Sigma-Aldrich

Fmoc-Ala-OH-13C3

99 atom % 13C, 99% (CP)

Synonym(s):

N-(9-Fluorenylmethoxycarbonyl)-L-alanine-13C3, L-Alanine-13C3, N-Fmoc

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$3,570.00

$3,570.00


Estimated to ship onMay 29, 2025


Request more information

Select a Size

Change View
1 G
$3,570.00

About This Item

Linear Formula:
13CH313CH(NH-Fmoc)13CO2H
Molecular Weight:
314.31
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$3,570.00


Estimated to ship onMay 29, 2025


Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

optical activity

[α]20/D -18°, c = 1 in DMF

technique(s)

bio NMR: suitable

mp

147-153 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

mass shift

M+3

SMILES string

[13CH3][13C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)[13C](O)=O

InChI

1S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16H,10H2,1H3,(H,19,22)(H,20,21)/t11-/m0/s1/i1+1,11+1,17+1

InChI key

QWXZOFZKSQXPDC-XZYMTLJFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service