Skip to Content
Merck
All Photos(1)

Key Documents

303240

Sigma-Aldrich

2-Methyl-3-buten-1-ol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$154.00

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$154.00

About This Item

Linear Formula:
H2C=CHCH(CH3)CH2OH
CAS Number:
Molecular Weight:
86.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$154.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.427 (lit.)

bp

120-121 °C/756 mmHg (lit.)

density

0.835 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(CO)C=C

InChI

1S/C5H10O/c1-3-5(2)4-6/h3,5-6H,1,4H2,2H3

InChI key

NVGOATMUHKIQQG-UHFFFAOYSA-N

General description

Secondary organic aerosol (SOA) formation from 2-methyl-3-buten-2-ol (MBO) photooxidation was investigated.[1]

Application

  • Semiochemical emission by fungal symbionts: The study explored how linoleic acid stimulates the emission of bark beetle semiochemicals, including compounds like 2-Methyl-3-buten-1-ol, by fungal symbionts, offering insights into ecological interactions and potential applications in pest management (Unelius et al., 2023).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F - closed cup

Flash Point(C)

33 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haofei Zhang et al.
Environmental science & technology letters, 1(4), 242-247 (2014-05-27)
Secondary organic aerosol (SOA) formation from 2-methyl-3-buten-2-ol (MBO) photooxidation has recently been observed in both field and laboratory studies. Similar to the level of isoprene, the level of MBO-derived SOA increases with elevated aerosol acidity in the absence of nitric
Tran B Nguyen et al.
Physical chemistry chemical physics : PCCP, 17(27), 17914-17926 (2015-06-23)
Methacryloyl peroxynitrate (MPAN), the acyl peroxynitrate of methacrolein, has been suggested to be an important secondary organic aerosol (SOA) precursor from isoprene oxidation. Yet, the mechanism by which MPAN produces SOA through reaction with the hydroxyl radical (OH) is unclear.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service