Skip to Content
Merck
All Photos(3)

Key Documents

129763

Sigma-Aldrich

2-Methyl-3-butyn-2-ol

98%

Synonym(s):

Dimethyl ethynyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC(CH3)2OH
CAS Number:
Molecular Weight:
84.12
Beilstein:
635746
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

15 mmHg ( 20 °C)

Quality Level

Assay

98%

form

liquid

autoignition temp.

662 °F

expl. lim.

16.6 %

refractive index

n20/D 1.42 (lit.)

bp

104 °C (lit.)

mp

2.6 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(C)(O)C#C

InChI

1S/C5H8O/c1-4-5(2,3)6/h1,6H,2-3H3

InChI key

CEBKHWWANWSNTI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-3-butyn-2-ol (MBY) is used as a precursor in the Mannich reaction and can undergo selective semihydrogenation to produce fine chemicals.[1][2]

Application

2-Methyl-3-butyn-2-ol can be used as a reactant to synthesize:,·
  • Aryl-2-methyl-3-butyn-2-ols via Pd-catalyzed Sonogashira coupling reaction with various aryl bromides.[3]      
  • 2-Methyl-3-buten-2-ol (MBE) by Pd/γ-Al2O3 catalyzed selective hydrogenation reaction. MBE is applicable as an important intermediate in the synthesis of vitamin A.[4]      
  • Diarylacetylenes via Pd-catalyzed Sonogashira coupling reaction with aryl chlorides in the presence of Cs2CO3 as a base.[5] 
  • Optically active propargylic alcohols by enantioselective addition reaction with various aldehydes in the presence of Zn(OTf)2 and N-methylephedrine.[6]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Palladium-Catalyzed Efficient and One-Pot Synthesis of Diarylacetylenes from the Reaction of Aryl Chlorides with 2-Methyl-3-butyn-2-ol
Yi Chenyi, et al.
Advanced Synthesis & Catalysis, 349(10), 1738-1742 (2007)
2-Methyl-3-butyn-2-ol as an acetylene precursor in the Mannich reaction. A new synthesis of suicide inactivators of monoamine oxidase
Fowler JS
The Journal of Organic Chemistry, 42(15) , 2637-2639 (1977)
Boyall et al.
Organic letters, 2(26), 4233-4236 (2001-01-11)
We report the first example of enantioselective aldehyde additions of 2-methyl-3-butyn-2-ol, a commodity bulk chemical that is readily available. Following a facile fragmentation reaction, the addition reactions provide access to optically active terminal acetylenes as useful building blocks for asymmetric
General kinetic modeling of the selective hydrogenation of 2-methyl-3-butyn-2-ol over a commercial palladium-based catalyst
Vernuccio S, et al.
Industrial & Engineering Chemistry Research, 54(46), 11543-11551 (2015)
Continuous 2-Methyl-3-Butyn-2-ol Selective Hydrogenation on Pd/?-Al2O3 as a Green Pathway of Vitamin A Precursor Synthesis
Fernandez-Ropero AJ, et al.
Catalysts, 11(4), 501-501 (2021)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service