Skip to Content
Merck
All Photos(1)

Key Documents

302368

Sigma-Aldrich

Acetoxyacetyl chloride

97%

Synonym(s):

α-Acetoxyacetyl chloride, (Acetyloxy)acetyl chloride, (Chlorocarbonyl)methyl acetate, 2-(Acetyloxy)acetyl chloride, 2-Acetoxyacetyl chloride, 2-Chloro-2-oxoethyl acetate, Acetic acid (chlorocarbonyl)methyl ester, Acetylglycoloyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$77.30
25 G
$251.00

$77.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$77.30
25 G
$251.00

About This Item

Linear Formula:
CH3CO2CH2COCl
CAS Number:
Molecular Weight:
136.53
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$77.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

55 °C/12 mmHg (lit.)

density

1.27 g/mL at 25 °C (lit.)

functional group

acyl chloride
ester

SMILES string

CC(=O)OCC(Cl)=O

InChI

1S/C4H5ClO3/c1-3(6)8-2-4(5)7/h2H2,1H3

InChI key

HZDNNJABYXNPPV-UHFFFAOYSA-N

Application

Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams[1] and glycolylhydroxamic acids.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naveen Anand et al.
The Journal of organic chemistry, 76(15), 5999-6006 (2011-06-15)
The facile synthesis of the stabilized axial and equatorial conformers of spiro-β-lactams was achieved via entrapment of cyclohexanone imines (Schiff bases) with acetoxyacetyl chloride in a [2 + 2]-cycloaddition reaction followed by their kinetic resolution. The immobilization of the racemic
M D Corbett et al.
Chemical research in toxicology, 1(4), 222-227 (1988-07-01)
A new and improved method for the synthesis of glycolylhydroxamic acids is described. The two-step method involves acylation of arylhydroxylamines with acetoxyacetyl chloride, followed by saponification of the ester bond to give the desired products. The conversion of hydroxamic acids

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service