Skip to Content
Merck
All Photos(2)

Key Documents

298328

Sigma-Aldrich

Lithium chloride hydrate

≥99.99% trace metals basis

Synonym(s):

Hydrochloric acid lithium salt hydrate, Lithium Chloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl · xH2O
CAS Number:
Molecular Weight:
42.39 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

Assay

≥99.99% trace metals basis

form

crystalline

impurities

≤100.0 ppm Trace Metal Analysis

SMILES string

[Li+].[Cl-].[H]O[H]

InChI

1S/ClH.Li.H2O/h1H;;1H2/q;+1;/p-1

InChI key

VXJIMUZIBHBWBV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boon-Hong Wee et al.
Nanoscale, 7(42), 17805-17811 (2015-10-13)
This article describes the effect of the lateral size of graphene oxide (GO) on the humidity sensing properties of a GO-based sensor. The GO size effect on the humidity sensing performance was evaluated on gold electrodes drop-coated with either an

Articles

Solid-state lithium fast-ion conductors are crucial for safer, high-energy-density all-solid-state batteries, addressing conventional battery limitations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service