Skip to Content
Merck
All Photos(2)

Documents

291838

Sigma-Aldrich

Propyl butyrate

99%

Synonym(s):

n-Propyl-n-butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2COOCH2CH2CH3
CAS Number:
Molecular Weight:
130.18
Beilstein:
1745552
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.4 (lit.)

bp

142-143 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

SMILES string

CCCOC(=O)CCC

InChI

1S/C7H14O2/c1-3-5-7(8)9-6-4-2/h3-6H2,1-2H3

InChI key

HUAZGNHGCJGYNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of thermal and nonthermal processing methods on apple cider quality and consumer acceptability.
Choi LH and Nielsen SS
Journal of Food Quality, 28(1), 13-29 (2005)
Jorge Montesinos et al.
Brain, behavior, and immunity, 45, 233-244 (2014-12-09)
The adolescent brain undergoes important dynamic and plastic cell changes, including overproduction of axons and synapses, followed by rapid pruning along with ongoing axon myelination. These developmental changes make the adolescent brain particularly vulnerable to neurotoxic and behavioral effects of
Amila M Devasurendra et al.
ACS applied materials & interfaces, 9(29), 24955-24963 (2017-07-05)
Pyrrole was covalently bonded to 1-methyl and 1-benzylimidazolium ionic liquids (ILs) via an N-substituted alkyl linkage to prepare electropolymerizable IL monomers with excellent thermal stability. The methylimidazolium IL, [pyrrole-C

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service